missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Tall-Form Beakers

Graduated to indicate approximate content

Manufacturer:  Corning Incorporated C1060400

 View more versions of this product

Catalog No. 02-546D



 

Specifications Description & Specifications

Specifications

Borosilicate Glass
Spout
2.71 in.
Yes
12.7cm
13.52 oz., 400mL
6.9cm
25mL
25 to 325mL
ASTM E960
Beaker
13.52 oz.
No
6 Pack
5 in.
Berzelius
White Enamel scale
Convenient for easy pouring
Marking spot
400mL
Accessories