missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Tall-Form Beakers

Graduated to indicate approximate content

Manufacturer:  Corning Incorporated C1060300

 View more versions of this product

Catalog No. 02-546C



 

Specifications Description & Specifications

Specifications

11.8cm
300mL
ASTM E960
6.4cm
Convenient for easy pouring
25 to 250mL
25mL
12 Pack
Berzelius
10.14 oz.
4.64 in.
Borosilicate Glass
2.51 in.
No
Yes
Spout
Marking spot
White Enamel scale
10.14 oz., 300mL
Beaker
Accessories