missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Tall-Form Beakers

Graduated to indicate approximate content

Manufacturer:  Corning Incorporated C1060100

 View more versions of this product

Catalog No. 02-546A



 

Specifications Description & Specifications

Specifications

7.9cm
100mL
ASTM E960
5cm
Convenient for easy pouring
20 to 80mL
10mL
12 Pack
Berzelius
3.38 oz.
3.11 in.
Borosilicate Glass
1.96 in.
No
Yes
Spout
Marking spot
White Enamel scale
3.38 oz., 100mL
Beaker
Accessories