Please login to your online account to display your discounted pricing
PYREX™ Straight Bore Stopcock with PTFE Product Standard Stopper Plug
plug stopcock taper PTFE plug fits into a microfinish barrel
$1,338.91 - $1,338.91
Specifications
Product Type Straight Bore Stopcock
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Side Arm Length (English) Plug Bore (English) Price Quantity    
14-629-58B
corning incorporated
C72824
0.39 in. 11.8 in. 0.157 in. Case of 6 Each for $1,338.91
Description & Specifications
Specifications
Product Type Straight Bore Stopcock
  • Freezing problems are reduced and greasing is eliminated
  • Very fine threads on the plug and nut let you “feel” the position of the plug
  • Side arm seals are heavy and strong for greater durability and easier handling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.