PYREX™ Stopcocks with Solid Glass standard taper Plug
For use in vacuum system to 1 × 10-4 torr
$1,058.64 - $1,265.68
Specifications
Product Type Stopcock
For Use With (Equipment) Vacuum system to 1 x 10-4 torr
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Side Arm Length (English) Plug Bore (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Side Arm Length (English) Plug Bore (English) Price Quantity  
14-560C Corning Incorporated
C72802
0.31 in. 9.8 in. 0.07 in. Case of 8 Each for $1,058.64
14-560E Corning Incorporated
C72804
0.39 in. 10 in. 0.157 in. Case of 8 Each for $1,265.68
Specifications
Description
  • Side arms, shell, and plug made of Pyrex brand tubing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Stopcock
Vacuum system to 1 x 10-4 torr