PYREX™ Squibb Separatory Funnels with joint (std. taper) Ground Joint and Glass Stopper
With standard taper stopper and stopcock
$347.17 - $445.45
Specifications
Material Pyrex™
For Use With (Application) For replacement glass standard taper stoppers, stopcock plugs and retaining clips
Joint Number 24/40
Quantity 1 Case
Stopper Material Glass
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outside Stopper Number Stopcock Number Price Quantity  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outside Stopper Number Stopcock Number Price Quantity  
10-438A Corning Incorporated
C6410125
4.22 oz. 2.37 in. 22 2 Case for $347.17
10-438B Corning Incorporated
C6410250
8.45 oz. 3.03 in. 22 2 Case for $384.43
10-438C Corning Incorporated
C6410500
16.9 oz. 3.8 in. 27 4 Case for $445.45
Specifications
Description
  • Drip tip in standard taper 24/40 joint at bottom
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Pyrex™
24/40
Glass
For replacement glass standard taper stoppers, stopcock plugs and retaining clips
1 Case