PYREX™ Shorty Disposable Glass Serological Pipets
Ideal for use in confined areas such as hoods
Manufacturer:  Corning Incorporated C7078B25
 View more versions of this product
Catalog No. 13-664-18D
Description
Description
  • Sterile, plugged pipets have tooled tops
  • Negative graduations make it applicable for both serological and measuring procedures
  • I.S.O. color coding is printed directly on Steri-View™ (one side paper, one side plastic) wrapper
Specifications
Specifications
Yes
0.84 oz.
Red
Yes
11.8 in.
5mL
Round
Serological Pipet
Pyrex™
Yes
0.84 oz., 25mL
Yes
0.2mL
300mm
Multi-pack
Tooled
25mL
Sterile
Documents
Accessories