PYREX™ Shorty Disposable Glass Serological Pipets
Ideal for use in confined areas such as hoods
Manufacturer:  Corning Incorporated C7077B10
 View more versions of this product
Catalog No. 13-676-29H
Description
Description
  • Sterile, plugged pipets have tooled tops
  • Negative graduations make it applicable for both serological and measuring procedures
  • I.S.O. color coding is printed directly on Steri-View™ (one side paper, one side plastic) wrapper
Specifications
Specifications
Yes
To Deliver/Blow Out
0.33 oz., 10mL
Yes
0.2mL
215mm
Individually Wrapped
Tooled
10mL
Sterile
Yes
0.33 oz.
Orange
Yes
8.4 in.
4mL
Round
Serological Pipet
Pyrex™
Documents
Accessories