missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Shaker Flasks with Extra-Deep Baffles

Long neck reduces splashing

Manufacturer:  Corning Incorporated C44463L

 View more versions of this product

Catalog No. 09-552-37


 

Specifications Description & Specifications

Specifications

Baffled
7.2 in.
185mm
13.26 in.
Delong
3000mL
Style: Shaker; Top style: Plain
3.8cm
Rotary or reciprocating shakers
33.7cm
Non-sterile
Borosilicate Glass
Accessories