Please login to your online account to display your discounted pricing

PYREX™ Reusable Media Storage Bottles


Ideal for media, buffers, aqueous and nonaqueous solutions

Manufacturer: corning incorporated  C1395150

 View more versions of this product

 Alert

Bottles larger than 2L should not have caps tightened immediately after autoclaving, nor should they be used with bottle-top filter units or other applications involving vacuum pressure, as breakage may occur.

Catalog No. 10-462-711

  • / Case of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Cs.
Capacity (Metric) 150mL
Capacity (English) 5.07 oz.
Closure Color Orange
Closure Screw
Closure Size GL45
Material Borosilicate Glass
Mouth Wide
Shape Cylindrical
Closure Material Polypropylene
Capacity 5.07 oz., 150mL