PYREX™ Reusable Class A Volumetric Serialized/Certified Pipets
Borosilicate-glass with fire-polished top and tip
Manufacturer:  Corning Incorporated C7101100
 View more versions of this product
Catalog No. 13-651-11K
Alert:
Do not pipet by mouth. Use mechanical pipetting device
Description
Description
Borosilicate-glass with fire-polished top and tip
PYREX Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory
Certifications
Each serialized pipet has a permanently marked serial number and Certificate of Identification and Capacity traceable to standards provided by NIST™.
Compliance
Calibrated “To Deliver” with tolerances in accordance with ASTM™ E 542 and E 969
Specifications
Specifications
To Deliver
A
Mechanical pipetting device
56.5cm
±0.08mL
100mL
Non-sterile
ASTM E542, ASTM E969, certificate of identification and capacity, NIST traceable
No
22.24 in.
Round
Serialized Volumetric Pipet
Pyrex™
Documents