Please login to your online account to display your discounted pricing
PYREX™ Replacement Tip For 2116-LSO
Manufacturer: corning incorporated  C2116GTO2
 View more versions of this product
Catalog No. 09-552-122
  • / Case of 12 Each
Description & Specifications
Specifications
For Use With (Equipment) 25mL and 50mL economy grade burets, used exclusively with the PTFE locking stopcock
Material Borosilicate Glass
Product Type Replacement Tip with ground and un-ground tip