PYREX™ PTFE Pestle Tissue Grinder with Steel Shaft
Recommended for soft tissues found in the liver and brain
Manufacturer:  Corning Incorporated C7724T50
 View more versions of this product
Catalog No. 11-850-60
Alert:
If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus from pieces of connective tissue or muscle. User should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass components.
Specifications Description & Specifications
Specifications
32mm
225mm
Borosilicate Glass
1.2 in.
1.69 oz., 30mL
30mL
Tissue Grinder
8.8 in.
1.69 oz.
Designed to handle both initial grinding and final homogenization in the same sequence of operations
PTFE threaded onto a stainless steel shaft
Pourout