missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Pharmaceutical Beakers

With double spout and double scale

Manufacturer:  Corning Incorporated C6480500

 View more versions of this product

Catalog No. 02-558C



 

Alert:

Do not subject directly to heat source.


Specifications Description & Specifications

Specifications

17.2cm
500mL
3.07 in.
No
Yes
Double spout
Marking spot
Dual white scale
16.9 oz.
6.77 in.
Borosilicate Glass
7.8cm
Ideal for measuring hot solutions
25 to 500mL
25mL
1 Pack
16.9 oz., 500mL
Beaker
Accessories