missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Pear-Shaped Flasks

With a single standard taper outer joint

Manufacturer:  Corning Incorporated C4315100

 View more versions of this product

Catalog No. 10-060-5E


 

Specifications Description & Specifications

Specifications

2.36 in.
No
11.5cm
14/20
230°C
Borosilicate Glass
60mm
4.5 in.
Short
446°F
100mL