PYREX™ Liebig Condensers, Standard Taper Outer and Inner Joints, Drip Tip
For vacuum distillation
Manufacturer:  PYREX™ C2400200
 View more versions of this product
Catalog No. 07-721A
Description
Description
For vacuum distillation
A general purpose condenser that can be used for distillation and extraction. Suitable for use in vacuum distillations. With the outer joint at the top and the inner joint at the bottom with a drip tip. The tubulation O.D. is approximately 10 mm.
Specifications
Specifications
For use in vacuum distillations
350 mm
200 mm
Glass
13.77 in.
7.87 in.
19/38
Liebig Condenser