Please login to your online account to display your discounted pricing
PYREX™ Large-Tip Opening Serological Pipets
Large-tip opening speeds intake and delivery of suspensions and viscous materials
$395.81 - $567.72
Specifications
Certifications/Compliance ASTM E1044
Material Pyrex™
Product Type Serological Pipet
Sterile No
Graduations 0.1mL
View More Specs
Products
Catalog Number Mfr. No. Accuracy Color Code Capacity (Metric) Price Quantity    
13-671-111E
corning incorporated
C708725
±0.20mL White 25mL Case of 12 Each for $567.72
13-671-111D
corning incorporated
C708710
±0.10mL Orange 10mL Case of 12 Each for $395.81
Description & Specifications
Specifications
Certifications/Compliance ASTM E1044
Material Pyrex™
Product Type Serological Pipet
Sterile No
Graduations 0.1mL
Tip Style Wide
Top Style Fire polished
  • Colored graduations in 0.01mL increments permanently enameled onto the glass
  • Fire-polished top and tip
  • Color coded by size for easy identification and sorting
Designed from ASTM™ E 1044. Calibrated “To Deliver/Blow Out”.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.