PYREX™ Large-Tip Opening Reusable Volumetric Pipets
Designed for schools and laboratories where Class A tolerances aren't required
Manufacturer:  Corning Incorporated C710220
 View more versions of this product
Catalog No. 13-650-6H
Description
Description
  • Larger tip openings result in faster deliveries, which in turn account for higher tolerances
  • Color coding on all but 50 and 100mL pipets permits easy sorting and identification by size
Compliance
Calibrated “To Deliver,” with tempered top and tip
Specifications
Specifications
To Deliver
Yellow
18.11 in.
Wide
Volumetric Pipet
Pyrex™
ASTM E969
No
46cm
±0.06mL
20mL
Non-sterile
Documents