PYREX™ Kjeldahl Flasks
Long necks, round bottoms, substantial walls
Manufacturer:  Corning Incorporated C5420100
 View more versions of this product
Catalog No. 10-110C
Specifications Description & Specifications
Specifications
Round
Top style: Threaded
5.5cm
8.26 in.
Long
Glass,Pyrex™
2
2.16 in.
No
21cm
100mL