PYREX™ Globe-Shaped Funnels with standard taper Stopcock and Stopper
Excellent for rack work
$223.98 - $461.23
Specifications
Closure Material Glass
For Use With (Application) For rack work
Includes Stopper and stopcock
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Stopcock Number Stopper Number Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Stopcock Number Stopper Number Height (English) Price Quantity  
10-442B Accessories Corning Incorporated
C6340125
4.22 oz. 2 22 12.9 in. Case for $223.98
10-442F Accessories Corning Incorporated
C63401L
33.8 oz. 4 27 16.2 in. Case for $461.23
10-442C Accessories Corning Incorporated
C6340250
8.4 oz. 2 22 14 in. Case for $261.57
Specifications
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Glass
For rack work
Stopper and stopcock