Please login to your online account to display your discounted pricing
PYREX™ Glass Manometers
Tube enclosed in round-bottom, graduated jacket and sealed to standard taper stopcock plug
$410.30 - $1,710.72
Products
Catalog Number Mfr. No. Price Quantity    
11-288
corning incorporated
C6950235
Case of 4 Each for $1,710.72
11-289
corning incorporated
C6952235
Case for $410.30
Description & Specifications
  • Compact
  • Read height of mercury column on 0 to 160mm scale graduated in 1mm divisions
  • Scale: 0 = 760mm (full vacuum); 80 = 680mm; 160 = 600mm
  • Without mercury
Not recommended for positive pressure
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.