missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Flasks with Short Ring Necks

Sometimes called “balloon flasks”

Manufacturer:  Corning Incorporated C42605L

 View more versions of this product

Catalog No. 10-065F



 

Specifications Description & Specifications

Specifications

Round
Top style: Ring
221mm
12.2 in.
Short
230°C
Borosilicate Glass
11
8.7 in.
No
31cm
446°F
5000mL
Accessories