PYREX™ Flash Chromatography Column, Spherical Joint Socket Joint, PTFE stopcock and 0-4mm PTFE valve
For quick separations in absorption chromatography applications
Manufacturer: Corning Incorporated C214945
Description
- 35/20 Spherical Joint socket joint at top and with 0-4mm PTFE plug stopcock at bottom
- Designed to be used with Flash reservoir (0572324 series) and flow controller
Specifications
Flash chromatography Column | |
1.77 in. | |
For Flash Reservoir and a Flow Controller | |
Pyrex Glass |
35/20 Spherical Joint socket joint at the top and with a 0 to 4 mm PTFE plug stopcock (at the bottom) | |
45 mm | |
450 mm | |
Straight Bore PTFE |