PYREX™ EZ Access™ Class A Low Actinic Flasks
Low actinic stained glass provides protection for materials sensitive to light
Manufacturer:  Corning Incorporated C55635100
 View more versions of this product
Catalog No. 13-646-22
Description
Description
  • Heavy-duty flasks, wide mouth facilitates filling and mixing
  • Easy-to-read graduation line is sharp and permanent
  • Class A tolerances
  • Retain mechanical strength, chemical stability and thermal resistance
Specifications
Specifications
Flat
16
65mm
Glass, Pyrex™
16cm
±0.05mL
Glass
2.5 in.
100mL
6.2 in.
Long
Documents