PYREX™ Evaporator Traps
Used when foam is likely to be created and drawn into the condenser of rotary evaporator
Manufacturer:  Corning Incorporated C400010014
 View more versions of this product
Catalog No. 09-549-1A
Specifications Description & Specifications
Specifications
3.4 oz., 100mL
15.8cm
6.25 in.
Rotary Evaporator Trap
2.4 in.
Rotary evaporator
3.4 oz.
Used when foam is likely to be created and drawn into the condenser of rotary evaporator
100mL
14/20 Inner Bottom, 24/40 Outer Top
6.2cm