PYREX™ Enlarging Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having larger size standard taper joint
Manufacturer:  Corning Incorporated C88202455
 View more versions of this product
Catalog No. 01-034G
Description
Description
  • Outer standard taper joint at top and larger inner standard taper joint at bottom
Specifications
Specifications
For attaching apparatus with one joint size to equipment having larger size joint
120mm
Tube Adapter
4.72 in.
Pyrex™
55/50 Inner Bottom, 24/40 Outer Top
Documents