PYREX™ Enlarging Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having larger size standard taper joint
Manufacturer: Corning Incorporated C88201024
Specifications
For attaching apparatus with one joint size to equipment having larger size joint | |
90mm | |
Tube Adapter |
3.54 in. | |
Pyrex™ | |
24/40 Inner Bottom, 10/30 Outer Top |