missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Dounce Tissue Grinder with Two Pestles

Two pestles dissociate cells into fine particles with minimal damage to cell nuclei

Manufacturer:  PYREX™ C77227

 View more versions of this product

Catalog No. 11-850-51



Includes: Unground glass tube (mortar) and two pestles

Description

Description

Supplied with two pestles to ensure dissociation of cells into fine particles with minimal damage to cell nuclei. Use the large clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided.

Specifications

Specifications

Non-sterile
7 mL
Dounce tissue grinder
Mortar tube, large pestle and small pestle
117 mm
Pourout
0.23 oz., 7 mL
0.23 oz.
Borosilicate Glass
Ideal for Enzyme studies where heat build-up must be avoided, use large clearance pestle for initial reduction of soft tissue, use small clearance pestle to complete homogenization
4.6 in.
0.59 in.
15 mm
Documents