Pyrex™ Round Bottom Threaded Culture Tubes
Made from borosilicate glass for use with samples sensitive to leaching from plastic tubes. Corning™ Pyrex™ Round Bottom Threaded Culture Tubes are designed for tissue culture and general bacteriological work.
Manufacturer:  Corning Incorporated C9944920
 View more versions of this product
Catalog No. 14-957-86E
Description
Description
  • Can be reliably used in almost any assay procedure from cell to chemical analysis
  • Made from chemically inert Pyrex™ borosilicate glass
  • Uniform base and wall thickness reduce the risk of breakage due to thermal shock and prevents warping or bending
  • Phenolic style screw caps available (separately) with PTFE liner for autoclave sterilisation, or rubber liner for hot air sterilisation
  • Supplied in convenient shelf pack quantities
Specifications
Specifications
Non-sterile
Screw neck
Yes
20mm
Borosilicate Glass
No
None
Round
18-415
24mL
20mm
Without
125mm
Yes
Ideal for use with samples sensitive to leaching from plastic tubes
Bulk
Untreated
Round
Documents
Accessories