PYREX™ Borosilicate Glass Flat Bottom Culture Tubes
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ PYREX™ Borosilicate Glass Flat Bottom Culture Tubes are designed for both tissue culture and general bacteriological work.
Manufacturer:  Corning Incorporated C9944816
 View more versions of this product
Catalog No. 14-957-86A
Description
Description
  • Not treated 
  • Threaded finish for screw cap
  • Disposable phenolic caps sold separately
  • Ideal for use with samples sensitive to leaching from plastic tubes
Specifications
Specifications
Non-sterile
Threaded
Yes
16mm
Borosilicate Glass
No
None
Flat
15-415
17mL
16mm
without
125mm
Yes
Ideal for use with samples sensitive to leaching from Plastic tubes
Bulk
Untreated
Flat
Documents
Accessories