Please login to your online account to display your discounted pricing
PYREX™ Disinfectant Cylinders
Designed for use in the Disinfectant Testing Procedure
$167.32 - $167.32
Specifications
Diameter (English) 0.31 in.
Diameter (Metric) 8mm
Material Borosilicate Glass
Product Type Disinfectant Testing Cylinder
For Use With (Application) For use in the disinfectant testing procedure as described in the 1992 J.A.O.A.C. hard surface carrier test for efficacy testing of disinfectants, collaborative study
View More Specs
Products
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity    
09-552-24
corning incorporated
C316710
0.39 in. 10mm Case of 125 Each for $167.32
Description & Specifications
Specifications
Diameter (English) 0.31 in.
Diameter (Metric) 8mm
Material Borosilicate Glass
Product Type Disinfectant Testing Cylinder
For Use With (Application) For use in the disinfectant testing procedure as described in the 1992 J.A.O.A.C. hard surface carrier test for efficacy testing of disinfectants, collaborative study
Sterile No
  • Use with procedure described in the 1992 J.A.O.A.C. (Journal for the Association of Official Analytical Chemists) Hard Surface Carrier Test for the Efficacy Testing of Disinfectants: Collaborative Study
  • Glass cylinders available in two sizes
  • Can be used in place of stainless-steel cylinders
  • Tubes are glazed on both ends