Please login to your online account to display your discounted pricing

PYREX™ Desiccator Replacement Covers


Various covers dependent on desiccator size and style

Manufacturer: corning incorporated  C3081150CO

 View more versions of this product

Catalog No. 08-624-412

  • / Case
 

Description & Specifications

Specifications

For Use With (Equipment) For small 2.4L knob nonvacuum desiccators
Height (Metric) 9.8cm
Height (English) 3.85 in.
Disposable No
Product Type Desiccator Cover

Constructed of Pyrex glass.