Please login to your online account to display your discounted pricing

PYREX™ Desiccator Replacement Covers


Various covers dependent on desiccator size and style

Manufacturer: corning incorporated  C3121150CO

 View more versions of this product

Catalog No. 08-624-421

  • / Case
 

Description & Specifications

Specifications

Height (Metric) 8.5cm
Disposable No
Product Type Desiccator Cover
For Use With (Equipment) Replacement for 2.4L small top vacuum desiccators
Height (English) 3.34 in.
Includes 24/29 standard taper stopcock