PYREX™ Cylindrical Tanks
Tops are ground to close tolerances to ensure tight cover fits
Manufacturer: PYREX™ C694536L
Description
A general-purpose jar with tops ground to close tolerances to insure a tight cover fit. The 152 mm jar edge is flat within 0.1 mm; all others are flat within 0.25 mm. Covers are not supplied. Do not use with heat, pressure, or vacuum applications.
Specifications
Cylindrical Chromatography Tank | |
Pyrex Glass | |
24 in. | |
60.9 cm | |
Ground | |
0.25 mm |
30.4 cm | |
12 in. | |
36 L | |
9.5 gal. | |
Cylindrical |