missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ PYREX™ Cylindrical Separatory Funnel, Graduated, PTFE Standard Stopcock, Glass Standard Taper Stopper
Microfinish PTFE stopcock plugs reduce freezing problems and eliminate contaminating greases
Supplier: Corning™ 6383250
Description
This funnel is graduated to show approximate capacity, has a micro-finish PTFE stopcock, and the strong, lightweight PYREX stopper.
For stopper only, see Cat. No. 7650
Specifications
52 mm | |
Pyrex Glass | |
Yes | |
2 mL | |
2 |
250 mL | |
340 mm | |
Standard stopcock and stopper | |
24/40 | |
Cylindrical Separatory Funnel |