missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Plain Cylindrical Jars

Can also be used as animal jars

Manufacturer:  PYREX™ C694232L

 View more versions of this product

Catalog No. 11-823M



Description

Description

These jars are mold-blown with substantial walls for mechanical strength. The open end of the jars is ground flat. These jars can also be used as animal jars. Do not use with heat, pressure, or vacuum applications.

Specifications

Specifications

406 mm
305 mm
1088 oz., 32000 mL
1088 oz.
Instrument Covers and Solution Containers
1/Cs.
Jar
12 in.
Round
32,000 mL
16 in.
Borosilicate Glass
1/Pk.
16 in.
SDS
Documents