missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PYREX™ Plain Cylindrical Jars
Can also be used as animal jars
Manufacturer: PYREX™ C69427L
Description
These jars are mold-blown with substantial walls for mechanical strength. The open end of the jars is ground flat. These jars can also be used as animal jars. Do not use with heat, pressure, or vacuum applications.
Specifications
222 mm | |
254 mm | |
256 oz., 7500 mL | |
256 oz. | |
Instrument Covers and Solution Containers | |
4/Cs. | |
Jar |
10 in. | |
Round | |
7500 mL | |
8.75 in. | |
Borosilicate Glass | |
1/Pk. | |
8.75 in. |