PYREX™ Culture Tubes with Threaded End and Marking Spot
For tissue culture and general bacteriological work
$551.60 - $959.74
Specifications
Design Type Threaded
Material Borosilicate Glass
Sterile No
Disposable Yes
For Use With (Application) For tissue culture work and general bacteriological use
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Thread Size Length (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Thread Size Length (Metric) Price Quantity  
14-962-110 Corning Incorporated
C99447161
11.5mL 16mm 15-415 100mm Case of 1000 Each for $882.13
14-962-111 Corning Incorporated
C9944720
24mL 20mm 18-415 125mm Case of 500 Each for $551.60
14-962-112 Corning Incorporated
C9944720X
30mL 20mm 18-415 150mm Case of 500 Each for $612.17
14-962C Corning Incorporated
C9944716X
19mL 16mm 15-415 150mm Case of 1000 Each for $959.74
14-962B Corning Incorporated
C9944716
15.5mL 16mm 15-415 125mm Case of 1000 Each for $944.74
14-962A Corning Incorporated
C9944713
7.5mL 13mm 13-415 100mm Case of 1000 Each for $828.60
Specifications
Description
  • For tissue culture and general bacteriological work
  • Borosilicate glass with round bottom
  • White marking spot
  • Threaded finish for screw cap (sold separately)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Threaded
No
For tissue culture work and general bacteriological use
White Marking Spot
Round
Borosilicate Glass
Yes
No
Bulk
Untreated