PYREX™ Culture Tubes with Threaded End and Marking Spot
For tissue culture and general bacteriological work
Manufacturer:  Corning Incorporated C9944716X
 View more versions of this product
Catalog No. 14-962C
Description
Description
  • For tissue culture and general bacteriological work
  • Borosilicate glass with round bottom
  • White marking spot
  • Threaded finish for screw cap (sold separately)
Specifications
Specifications
Non-sterile
Threaded
Without
Borosilicate Glass
No
White Marking Spot
Round
15-415
19mL
16mm
150mm
Yes
For tissue culture work and general bacteriological use
Bulk
Untreated
Documents