missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Crystallizing Dishes

Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization

Manufacturer:  Corning Incorporated C314090

 View more versions of this product

Catalog No. 08-741C



 

Specifications Description & Specifications

Specifications

No
270mL
90mm
9.13 oz., 270mL
Yes
3.54 in.
Crystallizing Dish
9.13 oz.
50mm
Ideal for storage and crystallization
Pyrex™
1.96 in.