missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Conical Reaction Vial

Replacement part for 6949M series of Pyrex microchemistry kits

Manufacturer:  Corning Incorporated C99005

 View more versions of this product

Catalog No. 03-338-10


 

Specifications Description & Specifications

Specifications

5.0mL
Reaction Vial
5mL
Screw
20mm
58mm
Conical
1.35 dr.