missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Class A Flasks with PTFE Stoppers

Manufacturer:  Corning Incorporated C5644200

 View more versions of this product

Catalog No. 10-462-859


 

Specifications

Description

Flask, Volumetric; Corning; PYREX; Class A; P.T.F.E. Stopper; Capacity: 200mL; Tolerance: ± 0.10mL; 74 x 219mm; Stopper No. 13

Constructed of sturdy Pyrex glass
Description & Specifications

Specifications

Flat
13
74mm
8.42 in.
Long
200mL
PTFE
2.9 in.
Yes
21.4cm
±0.1mL
Glass,Pyrex™