missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Class A Cylinders with Double Metric Scale

Calibrated “To Deliver”

Manufacturer:  Corning Incorporated C3023250

 View more versions of this product

Catalog No. 08-549-11D


 

Specifications Description & Specifications

Specifications

340mm
ASTM E1272, E542
3.9cm
250mL
Shock absorbing polyethylene bumper guards
±1mL
2mL
Borosilicate Glass
1.5 in.
A
13.3 in.
Double white metric scale
No
2mL
Accessories