Please login to your online account to display your discounted pricing
PYREX™ Chromatography Columns
Protects definition bands from mixing or smearing.
Manufacturer: fisher science education  3845210EMD
 View more versions of this product
Catalog No. S31154
  • / Pack of 2 Each
  • / Case of 2 Pack
Description & Specifications
Specifications
Packing Material With stopcock
Length (Metric) 300mm
Diameter (Metric) Inner 10.5mm
For Use With General chromatographic analysis
Type Chromatography Tube, 2/Pk.