Please login to your online account to display your discounted pricing
PYREX™ Chromatography Columns
Protects definition bands from mixing or smearing.
$713.30 - $713.30
Specifications
Diameter (Metric) Inner 10.5mm
Length (Metric) 300mm
For Use With General chromatographic analysis
View More Specs
Products
Catalog Number Mfr. No. Packing Material Type Price Quantity    
S31154
fisher science education
3845210EMD
With stopcock Chromatography Tube, 2/Pk. Pack of 2 Each for $713.30
Case of 2 Pack for $1,426.60
Description & Specifications
Specifications
Diameter (Metric) Inner 10.5mm
Length (Metric) 300mm
For Use With General chromatographic analysis
  • PYREX™ brand tube supports column packing with a sealed-in, coarse porosity, fritted disc
  • Tops tooled for rubber stopper fit
  • Without packing
  • Column without stopcocks has large diameter outflow tube that permits solvents to flow smoothly without smearing definition bands
  • Column with stopcock prevents definition bands from mixing by tapering the column beneath the disc