missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Polypropylene Caps for Erlenmeyer Flasks

As supplied with Pyrex™ Brand Media/Solution bottles. Choose orange caps for temperatures to 140°C (284°F), red caps for temperatures to 180°C (356°F).

Manufacturer:  PYREX™ C431340

 View more versions of this product

Catalog No. 07-202-530



Description

Description

Corning polypropylene Erlenmeyer flask caps are also available separately. These caps fit both Polycarbonate (PC) and Polyethylene terephthalate glycol-modified (PETG) Erlenmeyer Flasks and are available for the 500 mL and 1 L, 2L, 3L, and 5L flask sizes.

  • Sizes range from 500mL to 5L
  • Vent and flat cap options
  • Individually packaged
  • Sterility Assurance Level (SAL) of 10−6
  • RNase-/DNase-free
  • Nonpyrogenic
Specifications

Specifications

Vented
Sterile
Polypropylene
Style: Vented
3L Erlenmeyer flask
24/Cs.
Cap
140°C
Yes
Orange
Orange
7 cm
0.2μm pore nonwettable (hydrophobic) membrane
1/Pk.
284°F
SDS
Documents