Please login to your online account to display your discounted pricing

PYREX™ Capillary Tubing

Low-expansion, borosilicate glass tubing

$703.18 - $710.67

Products
Catalog Number Mfr. No. Price Quantity    

11-369-11D

  Accessories
corning incorporated
C236521
Case of 15 Each for $703.18

11-369-11C

  Accessories
corning incorporated
C236520
Case of 15 Each for $710.67
Description & Specifications

  • Softening point, 821°C (1509°F)
  • Working (forming) point, 1252°C (2885°F)

All tubing supplied in 4 ft. (1.2m) lengths.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.