missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ Burets with PYREX Stopcock

Class A

Manufacturer:  Corning Incorporated C213025

 View more versions of this product

Catalog No. 03-791-5



Includes: Carefully ground standard taper stopcock, 2mm bore stopcock plug, and dust cover.

Description

Description

  • Precision bore tubing assures accuracy between any two points
  • Durable colored graduations are sharply defined and easy to read

Compliance

Meet ASTM™ E 287 and E 42

Specifications

Specifications

0.84 oz.
A
1.2cm
75.5cm
Pyrex™
±0.03mL
0.84 oz., 25mL
0.1mL
ASTM E287, ASTM E542
0.47 in.
29.72 in.
Stopcock plug, dust cover
Pyrex™
25mL
Buret
Documents
Accessories