PYREX™ Buchner Funnels with Fritted Discs
Useful where filter paper would be attacked by material being filtered
Manufacturer:  Corning Incorporated C360603LC
 View more versions of this product
Catalog No. 10-358-2F
Specifications Description & Specifications
Specifications
101.44 oz.
ASTM E128
152mm
Fritted discs
101.44 oz., 3000mL
25mm
355mm
110mm
40 to 60μm
3000mL
5.98 in.
Useful where filter paper would be attacked by material being filtered
Coarse
0.98 in.
13.97 in.
4.33 in.
Pyrex™
Buchner Funnel