PYREX™ Biometer Culture Flask
Minimize the need for commonly used gas trains
$343.43 - $343.43
Specifications
Capacity (Metric) 25mL
View More Specs
Includes: Delivery funnel, #6 rubber stopper in the flask neck, #7 rubber stopper in the side tube, 16-gauge needle
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
11-850-15 Corning Incorporated
C4443250
25mL Case for $343.43
Specifications
Description
  • Measure production of carbon dioxide produced by microorganisms grown on a variety of culture media over longer periods of time
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
25mL